accessibility menu, dialog, popup

UserName

OHOClFFF(E/Z)

CAS RN 1092460-80-4

IUPAC Name: 3-[3-chloro-4-(trifluoromethyl)phenyl]prop-2-enoic acid
Synonyms: 3-[3-chloro-4-(trifluoromethyl)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 250.6
Molecular Formula: C10H6ClF3O2
InChi Key: JERIPOGIKJJMJU-UHFFFAOYSA-N
SMILES: OC(=O)C=CC1=CC(Cl)=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results