accessibility menu, dialog, popup

UserName

NClFFF

CAS RN 1092460-79-1

IUPAC Name: 3-chloro-4-(trifluoromethyl)benzonitrile
Synonyms: 3-chloro-4-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 205.56
Molecular Formula: C8H3ClF3N
InChi Key: IWZFPSVQIDJRAD-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(Cl)C=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results