CAS RN 1084-32-8
CAS RN 1084-32-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5,7-Dinitro-8-quinolinol, TRC
CAS: 1084-32-8 Molecular Formula: C9H5N3O5 Molecular Weight (g/mol): 235.15 Synonym: 5,7-Bis(hydroxyl[oxido]amino)-8-hydroxyquinoline,5,7-Dinitro-8-hydroxyquinoline,5,7-Dinitrooxine,8-Hydroxy-5,7-dinitroquinoline,NSC 57103,NSC 74940 IUPAC Name: 5,7-dinitroquinolin-8-ol SMILES: OC1=C2C(C=CC=N2)=C([N+]([O-])=O)C=C1[N+]([O-])=O