accessibility menu, dialog, popup

UserName

CAS RN 1073371-78-4

BOOH3CCH3H3CH3CNClCl

CAS RN 1073371-78-4

IUPAC Name: 2,6-dichloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2,6-dichloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 273.95
Molecular Formula: C11H14BCl2NO2
InChi Key: AOLXLWZJOQBPQX-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(Cl)N=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results