CAS RN 1071-28-9
CAS RN 1071-28-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
3-Aminopropyl Monophosphate, TRC
CAS: 1071-28-9 Molecular Formula: C3 H10 N O4 P Molecular Weight (g/mol): 155.09 Synonym: 1-Propanol, 3-amino-, 1-(dihydrogen phosphate),1-Propanol, 3-amino-, dihydrogen phosphate (ester) (6CI,7CI,8CI,9CI),3-Aminopropyl dihydrogen phosphate,3-Aminopropyl monophosphate IUPAC Name: 3-aminopropyl dihydrogen phosphate SMILES: NCCCOP(=O)(O)O
2
Cyclophosphamide EP Impurity F (Cyclophosphamide USP Related Compound C), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.