accessibility menu, dialog, popup

UserName

CAS RN 107007-99-8

IUPAC Name: hydrogen 1-methyl-N-{9-methyl-9-azabicyclo[3.3.1]nonan-3-yl}-1H-indazole-3-carboxamide chloride
Synonyms: granisetron
Molecular Weight (g/mol): 348.88
Molecular Formula: C18H25ClN4O
InChi Key: QYZRTBKYBJRGJB-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CN1N=C(C(=O)NC2CC3CCCC(C2)N3C)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (7)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)