CAS RN 106261-48-7
IUPAC Name:
4-[(4-methylpiperazin-1-yl)methyl]benzoic acid
Synonyms:
4-[(4-methylpiperazin-1-yl)methyl]benzoic acid
Molecular Weight (g/mol):
234.3
Molecular Formula:
C13H18N2O2
InChi Key:
ZJUXJQSYXBYFFO-UHFFFAOYSA-N
SMILES:
CN1CCN(CC2=CC=C(C=C2)C(O)=O)CC1
1
–
4
of
4
results
1
4-(4-Methyl-1-piperazinylmethyl)benzoic acid, 97%, Thermo Scientific™
CAS: 106261-48-7 Molecular Formula: C13H18N2O2 Molecular Weight (g/mol): 234.299 MDL Number: MFCD01632530 InChI Key: ZJUXJQSYXBYFFO-UHFFFAOYSA-N PubChem CID: 4769782 IUPAC Name: 4-[(4-methylpiperazin-1-yl)methyl]benzoic acid SMILES: CN1CCN(CC1)CC2=CC=C(C=C2)C(=O)O
2
4-(4-Methylpiperazin-1-ylmethyl)benzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-(4-Methylpiperazin-1-ylmethyl)benzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
4-(4-Methylpiperazin-1-ylmethyl)benzoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.