missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methylpiperazin-1-ylmethyl)benzoic Acid, TRC

Product Code. 30471061
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30471061 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471061 Supplier Toronto Research Chemicals Supplier No. TRCM2193501G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(4-Methylpiperazin-1-ylmethyl)benzoic Acid
CAS 106261-48-7
Molecular Formula C13 H18 N2 O2
Quantity 1 g
Synonym 4-[(4-Methyl-1-piperazinyl)methyl]benzoic Acid, 4-((4-Methylpiperazin-1-yl)methyl)benzoic acid, 4-(4-Methylpiperazinyl)methylbenzoic acid
InChI Formula InChI=1S/C13H18N2O2/c1-14-6-8-15(9-7-14)10-11-2-4-12(5-3-11)13(16)17/h2-5H,6-10H2,1H3,(H,16,17)
SMILES CN1CCN(Cc2ccc(cc2)C(=O)O)CC1
IUPAC Name 4-[(4-methylpiperazin-1-yl)methyl]benzoic acid
Molecular Weight (g/mol) 234.294
Formula Weight 234.137
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.