CAS RN 1044904-10-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Topotecan-d6, TRC
CAS: 1044904-10-0 Molecular Formula: C23 2H6 H17 N3 O5 Molecular Weight (g/mol): 427.48 Synonym: 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 10-[[di(methyl-d3)amino]methyl]-4-ethyl-4,9-dihydroxy-, (4S)-,Topotecan-d6,Topotecan D6 (N,N-dimethyl D6) SMILES: [2H]C([2H])([2H])N(Cc1c(O)ccc2nc3C4=CC5=C(COC(=O)[C@]5(O)CC)C(=O)N4Cc3cc12)C([2H])([2H])[2H]