accessibility menu, dialog, popup

UserName

CAS RN 102783-51-7

ONNONH2OPOOHOPOOHOPOHOOHOHNaNa

CAS RN 102783-51-7

IUPAC Name: ({[({[5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy](hydroxy)phosphoryl}oxy)phosphonic acid disodium
Synonyms: deoxycytidine triphosphate disodium
Molecular Weight (g/mol): 513.14
Molecular Formula: C9H16N3Na2O13P3
InChi Key: QGRIWAGEUGZHQH-UHFFFAOYNA-N
SMILES: [Na].[Na].NC1=NC(=O)N(C=C1)C1CC(O)C(COP(O)(=O)OP(O)(=O)OP(O)(O)=O)O1
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)