missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxycytidine-5'-Triphosphate Trisodium Salt, >98%, MP Biomedicals™

Product Code. 11487640 Shop All MP Biomedicals Products
Click to view available options
:
500mg
This item is not returnable. View return policy

Product Code. 11487640

Brand: MP Biomedicals 0210055890

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2'-Deoxycytidine 5'-triphosphate (dCTP) is used to synthesize DNA via various DNA polymerases or reverse transcriptases. dCTP is used in studies of nucleotide pools, nucleotide selectivity and enzyme specificity studies

TRUSTED_SUSTAINABILITY

Specifications

CAS 102783-51-7
Assay Percent Range >98%
Molecular Formula C9H16N3Na2O13P3
MDL Number MFCD00084683,MFCD00080336
Quantity 500 mg
Synonym dCTP, 2'-Deoxycytidine 5'-triphosphate disodium salt
InChI Key QGRIWAGEUGZHQH-UHFFFAOYNA-N
SMILES [Na].[Na].NC1=NC(=O)N(C=C1)C1CC(O)C(COP(O)(=O)OP(O)(=O)OP(O)(O)=O)O1
IUPAC Name ({[({[5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy](hydroxy)phosphoryl}oxy)phosphonic acid disodium
Molecular Weight (g/mol) 513.14
PubChem CID 131636617
Formula Weight 533.11
Physical Form Solid
Chemical Name or Material 2'-Deoxycytidine-5'-Triphosphate Trisodium Salt
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.