CAS RN 102690-88-0
CAS RN 102690-88-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Bis(2-cyanoethyl)-N,N-diisopropyl Phosphoramidite, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Bis(2-cyanoethyl)-N,N-diisopropyl Phosphoramidite, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Bis(2-cyanoethyl)-N,N-diisopropyl Phosphoramidite, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.