Learn More
Bis(2-cyanoethyl)-N,N-diisopropyl Phosphoramidite, TRC
Specifications
Specifications
| Chemical Name or Material | Bis(2-cyanoethyl)-N,N-diisopropyl Phosphoramidite |
| CAS | 102690-88-0 |
| Molecular Formula | C12 H22 N3 O2 P |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C12H22N3O2P/c1-11(2)15(12(3)4)18(16-9-5-7-13)17-10-6-8-14/h11-12H,5-6,9-10H2,1-4H3 |
| SMILES | CC(C)N(C(C)C)P(OCCC#N)OCCC#N |
| IUPAC Name | 3-[2-cyanoethoxy-[di(propan-2-yl)amino]phosphanyl]oxypropanenitrile |
| Molecular Weight (g/mol) | 271.3 |
| Formula Weight | 271.145 |