CAS RN 1025814-13-4
CAS RN 1025814-13-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
N-(2-Chloro-6-methylphenyl)-2-[[6-[[2-[(2-hydroxyethyl)amino]ethyl]amino]-2-methyl-4-pyrimidinyl]amino]- 5-thiazolecarboxamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-(2-Chloro-6-methylphenyl)-2-[[6-[[2-[(2-hydroxyethyl)amino]ethyl]amino]-2-methyl-4-pyrimidinyl]amino]- 5-thiazolecarboxamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-(2-Chloro-6-methylphenyl)-2-[[6-[[2-[(2-hydroxyethyl)amino]ethyl]amino]-2-methyl-4-pyrimidinyl]amino]- 5-thiazolecarboxamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.