Learn More
N-(2-Chloro-6-methylphenyl)-2-[[6-[[2-[(2-hydroxyethyl)amino]ethyl]amino]-2-methyl-4-pyrimidinyl]amino]- 5-thiazolecarboxamide, TRC
Specifications
Specifications
| Chemical Name or Material | N-(2-Chloro-6-methylphenyl)-2-((6-((2-((2-hydroxyethyl)amino)ethyl)amino)-2-methyl-4-pyrimidinyl)amino)- 5-thiazolecarboxamide |
| CAS | 1025814-13-4 |
| Molecular Formula | C20 H24 Cl N7 O2 S |
| Quantity | 25 mg |
| Synonym | N-(2-Chloro-6-methylphenyl)-2-[[6-[[2-[(2-hydroxyethyl)amino]ethyl]amino]-2-methyl-4-pyrimidinyl]amino]- 5-thiazolecarboxamide, 5-Thiazolecarboxamide, N-(2-chloro-6-methylphenyl)-2-[[6-[[2-[(2-hydroxyethyl)amino]ethyl]amino]-2-methyl-4-pyrimidinyl]amino]- |
| InChI Formula | InChI=1S/C20H24ClN7O2S/c1-12-4-3-5-14(21)18(12)28-19(30)15-11-24-20(31-15)27-17-10-16(25-13(2)26-17)23-7-6-22-8-9-29/h3-5,10-11,22,29H,6-9H2,1-2H3,(H,28,30)(H2,23,24,25,26,27) |
| SMILES | Cc1nc(NCCNCCO)cc(Nc2ncc(s2)C(=O)Nc3c(C)cccc3Cl)n1 |
| IUPAC Name | N-(2-chloro-6-methylphenyl)-2-[[6-[2-(2-hydroxyethylamino)ethylamino]-2-methylpyrimidin-4-yl]amino]-1,3-thiazole-5-carboxamide |
| Molecular Weight (g/mol) | 461.968 |
| Formula Weight | 461.14 |