CAS RN 1025718-91-5
CAS RN 1025718-91-5
IUPAC Name:
methyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carboxylate
Synonyms:
methyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carboxylate
Molecular Weight (g/mol):
263.1
Molecular Formula:
C13H18BNO4
InChi Key:
FJOJOCXHLGXEQA-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CN=CC(=C1)B1OC(C)(C)C(C)(C)O1
1
–
2
of
2
results
1
3-(Methoxycarbonyl)pyridine-5-boronic acid, pinacol ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-(Methoxycarbonyl)pyridine-5-boronic acid, pinacol ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.