Learn More
3-(Methoxycarbonyl)pyridine-5-boronic acid, pinacol ester, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(Methoxycarbonyl)pyridine-5-boronic acid, pinacol ester |
| CAS | 1025718-91-5 |
| Molecular Formula | C13H18BNO4 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C13H18BNO4/c1-12(2)13(3,4)19-14(18-12)10-6-9(7-15-8-10)11(16)17-5/h6-8H,1-5H3 |
| SMILES | O=C(C1=CC(B2OC(C)(C(C)(C)O2)C)=CN=C1)OC |
| IUPAC Name | methyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinate |
| Molecular Weight (g/mol) | 263.1 |
| Formula Weight | 263.13 |