accessibility menu, dialog, popup

UserName

CAS RN 10252-29-6

OClNNOCl(E/Z)

CAS RN 10252-29-6

IUPAC Name: 4-{2-[4-(carbonochloridoyl)phenyl]diazen-1-yl}benzoyl chloride
Synonyms: 4-{2-[4-(carbonochloridoyl)phenyl]diazen-1-yl}benzoyl chloride
Molecular Weight (g/mol): 307.13
Molecular Formula: C14H8Cl2N2O2
InChi Key: ASOXKYGOZZTVHL-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)C(Cl)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)