Learn More
Azobenzene-4,4'-dicarbonyl Dichloride, TRC
Specifications
Specifications
| Chemical Name or Material | Azobenzene-4,4'-dicarbonyl Dichloride |
| CAS | 10252-29-6 |
| Molecular Formula | C14H8Cl2N2O2 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C14H8Cl2N2O2/c15-13(19)9-1-5-11(6-2-9)17-18-12-7-3-10(4-8-12)14(16)20/h1-8H |
| SMILES | ClC(=O)c1ccc(cc1)N=Nc2ccc(cc2)C(=O)Cl |
| IUPAC Name | 4-[(E)-(4-carbonochloridoylphenyl)diazenyl]benzoyl chloride |
| Molecular Weight (g/mol) | 307.13 |
| Formula Weight | 305.9963 |