CAS RN 10238-27-4
CAS RN 10238-27-4
IUPAC Name:
(2S,3R,4S,5R)-2-(2-nitrophenoxy)oxane-3,4,5-triol
Synonyms:
(2S,3R,4S,5R)-2-(2-nitrophenoxy)oxane-3,4,5-triol
Molecular Weight (g/mol):
271.23
Molecular Formula:
C11H13NO7
InChi Key:
YPQCLGUTGDQYNI-DQDDRIPDSA-N
SMILES:
O[C@@H]1CO[C@@H](OC2=CC=CC=C2[N+]([O-])=O)[C@H](O)[C@H]1O
1
–
3
of
3
results
1
2’-Nitrophenyl-Beta-D-xylopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2’-Nitrophenyl-Beta-D-xylopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2’-Nitrophenyl-Beta-D-xylopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.