Learn More
2’-Nitrophenyl-Beta-D-xylopyranoside, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Nitrophenyl-beta-D-xylopyranoside |
| CAS | 10238-27-4 |
| Molecular Formula | C11 H13 N O7 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C11H13NO7/c13-7-5-18-11(10(15)9(7)14)19-8-4-2-1-3-6(8)12(16)17/h1-4,7,9-11,13-15H,5H2/t7-,9+,10-,11+/m1/s1 |
| SMILES | O[C@@H]1CO[C@@H](Oc2ccccc2[N+](=O)[O-])[C@H](O)[C@H]1O |
| IUPAC Name | (2S,3R,4S,5R)-2-(2-nitrophenoxy)oxane-3,4,5-triol |
| Molecular Weight (g/mol) | 271.22 |
| Formula Weight | 271.0692 |