CAS RN 1018899-04-1
CAS RN 1018899-04-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sotagliflozin, MedChemExpress
MedChemExpress Sotagliflozin (LX-4211) is a potent dual SGLT2/1 inhibitor. Antidiabetic agents.