missing translation for 'onlineSavingsMsg'
Learn More

Sotagliflozin, MedChemExpress

Product Code. 16489157
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16489157 5 mg 5mg
16403486 25 mg 25mg
16412687 50 mg 50mg
16486588 10 mg 10mg
4 options
This item is not returnable. View return policy

Product Code. 16489157

Brand: Medchem Express HY15516

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sotagliflozin (LX-4211) is a potent dual SGLT2/1 inhibitor. Antidiabetic agents.

Sotagliflozin (LX-4211) is a potent dual SGLT2/1 inhibitor. Antidiabetic agents.

Specifications

Chemical Name or Material Sotagliflozin
CAS 1018899-04-1
Color White
Purity Grade Notes Research
Molecular Formula C21H25ClO5S
Quantity 5 mg
Synonym LX-4211 LP-802034
Solubility Information DMSO : ≥ 100 mg/mL (235.33 mM)
SMILES ClC(C=CC([C@@H]1O[C@H](SC)[C@@H](O)[C@H](O)[C@H]1O)=C2)=C2CC3=CC=C(OCC)C=C3
Molecular Weight (g/mol) 424.94
Formula Weight 424.94
Percent Purity 98.77%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.