CAS RN 1017779-68-8
IUPAC Name:
1-(4-ethoxy-2,3-difluorophenyl)ethan-1-one
Synonyms:
1-(4-ethoxy-2,3-difluorophenyl)ethanone
Molecular Weight (g/mol):
200.19
Molecular Formula:
C10H10F2O2
InChi Key:
YHGFVKNXGQWJMM-UHFFFAOYSA-N
SMILES:
CCOC1=C(F)C(F)=C(C=C1)C(C)=O
1
–
1
of
1
results
1
4'-Ethoxy-2',3'-difluoroacetophenone, 97%, Thermo Scientific™
CAS: 1017779-68-8 Molecular Formula: C10H10F2O2 Molecular Weight (g/mol): 200.185 MDL Number: MFCD09258710 InChI Key: YHGFVKNXGQWJMM-UHFFFAOYSA-N Synonym: 4'-ethoxy-2',3'-difluoroacetophenone,1-4-ethoxy-2,3-difluorophenyl ethanone,2',3'-difluoro-4'-ethoxyacetophenone,1-acetyl-4-ethoxy-2,3-difluorobenzene,4'-ethoxy-2',3'-difluoroacetophenone, jrd PubChem CID: 46737625 IUPAC Name: 1-(4-ethoxy-2,3-difluorophenyl)ethanone SMILES: CCOC1=C(C(=C(C=C1)C(=O)C)F)F