accessibility menu, dialog, popup

UserName

CAS RN 1017777-95-5

IUPAC Name: 3-[2-methyl-3-(trifluoromethyl)phenyl]prop-2-enoic acid
Synonyms: 3-[2-methyl-3-(trifluoromethyl)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 230.19
Molecular Formula: C11H9F3O2
InChi Key: UDOYNGWYVWNFOC-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(O)=O)C=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results