accessibility menu, dialog, popup

UserName

CH3FFFN

CAS RN 1000515-00-3

IUPAC Name: 2-[2-methyl-3-(trifluoromethyl)phenyl]acetonitrile
Synonyms: 2-[2-methyl-3-(trifluoromethyl)phenyl]acetonitrile
Molecular Weight (g/mol): 199.18
Molecular Formula: C10H8F3N
InChi Key: VSBABDIMZJXXIU-UHFFFAOYSA-N
SMILES: CC1=C(CC#N)C=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results