missing translation for 'onlineSavingsMsg'
Learn More

ZsGreen1 Antibody (OTI3C4), Novus Biologicals™

Product Code. 18688870 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18688870 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18688870 Supplier Novus Biologicals Supplier No. NBP278867

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZsGreen1 Monoclonal antibody specifically detects ZsGreen1 in Nonspecies specific samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZsGreen1
Applications Western Blot
Classification Monoclonal
Clone OTI3C4
Concentration 3.19 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000
Formulation PBS (pH 7.3), 1% BSA, 50% Glycerol
Gene Alias zsgreen
Host Species Mouse
Immunogen A ZsGreen1 tagged recombinant protein expressed in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at -20°C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.