Learn More
Invitrogen™ ZOOM™ Equilibration Trays
Product Code. 11580906
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11580906 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11580906 Supplier Invitrogen™ Supplier No. ZM0007
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Convenient, disposable trays
ZOOM Equilibration Trays are designed for use with the ZOOM IPGRunner Cassettes to perform equilibration of ZOOM Strips after first-dimension isoelectric focusing and prior to second-dimension (2D) SDS-PAGE.
  • Simplify the equilibration and incubation process in 2D electrophoresis
  • Savings in time and labor
  • Optimal equilibration of samples
  • Minimal risk of sample contamination and damage to IPG strips
2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room temperature
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage The ZOOM™ Equilibration Trays are supplied in a pack of 10. The combo kit of ZOOM™Cassettes and ZOOM™ Equilibration Trays includes a box of 10 cassettes and a pack of 10 trays. Store at room temperature. Guaranteed stable for one year when properly stored.
Quantity 10/Pk.
Shipping Condition Room Temperature
Product Line ZOOM
Product Type Equilibration Tray
For Use With (Equipment) ZOOM IPGRunner Mini
I just set up the ZOOM IPGRunner for IEF but don't see any current running through the system. What is the issue?
A possible reason is poor contact between electrodes or incomplete circuit. Make sure that you have added 600 µL deionized water to the Electrode Wicks and the gel is exposed at the anodic and cathodic ends of the cassette. Check the power supply. Be sure to set the “Load Check” to off to enable the power supply to operate at low current.
What are the components of the ZOOM IPGRunner Mini-Cell?
Here are the components of the ZOOM IPGRunner Mini-Cell:
ZOOM IPGRunner Core
ZOOM IPGRunner Lid
Gel Tension Wedge
Buffer Dam
Mini-Cell Chamber
I used the ZOOM IPGRunner system and some of my protein spots are missing or appear as a smear. Can you please offer some suggestions?
Here are the possible causes and solutions:
*Protein degradation: Add protease inhibitors during sample preparation (see manual for details).
*Different subunits: Use denaturing conditions (8 M urea).
*Incomplete equilibration: Perform equilibration as described in the manual. Increase the equilibration time.
*Protein precipitates: Increase solubilization reagents in the rehydration buffer (see manual for details. Use appropriate strips based on the pI of the protein sample.
*Low protein load: Increase the protein load. You can load up to 400 µg of fractionated protein sample per ZOOM Strip. Use an accurate and sensitive protein estimation method.
*Insensitive detection method: Use sensitive detection methods such as silver staining or immunoblotting.
I used the ZOOM IPGRunner system and got vertical streaking. Can you please offer some suggestions?
Here are the possible causes and solutions:
*Protein has got oxidized: Include DTT in the rehydration buffer and perform the alkylation step.
*Impure solutions: Use ultrapure reagents to prepare the rehydration buffer, equilibration buffer, and buffers for SDS/PAGE.
I used the ZOOM IPGRunner System and got horizontal streaking. Can you please offer some suggestions?
Here are the possible causes and solutions:
*Impure solutions: Use ultrapure reagents to prepare the rehydration buffer, equilibration buffer, and buffers for SDS/PAGE.
*Air bubble between the strip and 2D gel: Smooth out the air bubbles.
*Poor strip rehydration: Rehydrate the strips in 140 µL rehydrating buffer for 1 hour as described in the manual. Rehydration can be extended to overnight if you use 155 µL rehydrating buffer. Make sure that the rehydration buffer covers the strip completely.
*Protein overload: Decrease the protein concentration or lower the sample volume.
*Protein precipitates: Increase solubilization reagents in the rehydration buffer (see manual). Use appropriate strips based on the pI of the protein sample. Do not add more than 10 µL of your sample to 140 µL of rehydration buffer (see manual).
I used the ZOOM IPGRunner System and got incomplete focusing of my proteins. Can you please offer some tips?
Here are the possible causes and solutions:
*Improper sample preparation: Increase solubilization reagents in the rehydration buffer. Use 8 M urea for solubilization. Add DTT and non-ionic detergents as described in the manual.
*Incorrect focusing time: Increase or decrease the focusing time based on the initial results.
*High protein load: Decrease the protein load. Use an accurate and sensitive protein estimation method.
*Salt, lipids, or nucleic acid impurities present in the sample: Remove interfering substances (see manual for details).
*Poor strip rehydration: Rehydrate the strips in 140 µL rehydrating buffer for 1 hour as described in the manual. Rehydration can be extended to overnight if you use 155 µL rehydrating buffer. Make sure the rehydration buffer is covering the strip completely.
After the first dimension separation using the ZOOM IPGRunner System, I do not see any distinct spots. What could the issue be?
Here are the possible causes and solutions:
*Low protein load. Increase the protein load. Use an accurate and sensitive protein estimation method.
*Improper sample preparation. Increase solubilization reagents. Use at least 8 M urea for solubilization. Add DTT and non-ionic detergents (see manual for details).
*Strip not correctly oriented. Align the strip correctly as described in the manual. Be sure to have the gel side up when loading the strip into the ZOOM IPGRunner Cassette.
*Air bubbles between the strip and 2D gel. Smooth out any air bubbles.
*Insensitive detection method. Use sensitive detection methods such as silver staining or immunoblotting.
What are the power supply considerations for the ZOOM IPGRunner system?
The ZOOM IPGRunner System should be used with an external DC power supply designed for IEF and electrophoresis applications. This power supply must:
*Be isolated from the ground so that the DC output is floating.
*Be programmable, with a 4-protocol minimum.
*Be able to operate at low current (<1 mA) as IEF is performed at very low current.
Note: Many power supplies automatically shut off when the current drops below 1 mA. You will need a power supply capable of overriding the low current shut-off feature.
The electrical leads of the ZOOM IPGRunner Lid are recessed and may not fit into some power supply units. To allow connection of the ZOOM IPGRunner power leads with certain power supplies, use Invitrogen Power Supply Adapters available separately (Cat. No. ZA10001).
What is the optimal concentration of detergent that I should add to my sample for the ZOOM IPG Runner?
Detergents are used in the sample as solubilizing agents:
*SDS must be 0.2% or lower. If the percentage of SDS is too high, all the proteins will be negatively charged and will migrate to the positive electrode.
*CHAPS can be increased up to 4%.
*NP-40, Triton-X and other nonionic detergents are used at 0.5-0.7%.
What is the optimal salt concentration that I should use in my sample for the ZOOM IPG Runner?
The salt concentration should not exceed 10 mM for these reasons:
*Burning can occur when conductivity is high, hence current limits should be set.
*Protein transport in an electric field decreases with increasing ionic strength.
*The efficiency of all IEF instruments is decreased when samples contain more than 10 mM salt.
*Longer run times are generally required for samples containing >10 mM salt.
Can I run non-reduced samples in the ZOOM IPGRunner?
You can run non-reduced samples in the ZOOM IPGRunner by omitting DTT in the rehydration buffer and equilibration buffers.
Can I heat my protein sample prepared using ZOOM 2D Protein Solubilizer?
We do not recommend heating protein samples containing urea over 37 degrees C as elevated temperatures cause urea to hydrolyze to isocyanate, which modifies proteins by carbamylation.
How do you recommend storing protein samples prepared in the Sample Rehydration buffer?
We recommend storing them at -80 degrees C. We do not recommend storing them at -20 degrees C
How much ampholyte do you recommend adding to the sample rehydration buffer?
The recommended ampholyte concentration in the sample rehydration buffer is 0.5%.
*If you are loading 5-50 µg of protein (pure protein or crude lysate) per ZOOM strip, use 0.5% ampholytes in the sample rehydration buffer.
*If you are loading >50 µg of protein (crude lysate or fractionated sample) per ZOOM Strip, use 0.5-2% ampholytes in the sample rehydration buffer.
Note: Higher ampholyte concentration requires longer focusing times.
What is the maximum recommended volume of protein sample to be added to the Sample Rehydration buffer?
The maximum volume of the protein sample should at most be 1/6 of the final sample volume that will be added to the strip. A good general volume would be 5-10µL. 140 µL of sample diluted in Sample Rehydration buffer is used to rehydrate each ZOOM Strip for the standard rehydration time of one hour. For overnight rehydration, we recommend using 155 µL of diluted sample.
I am using your ZOOM IPGRunner system. What is the sample rehydration buffer?
The sample rehydration buffer, also known as the sample buffer, is used to denature and solubilize protein samples, and rehydrate the IPG strips prior to performing IEF using the ZOOM IPGRunner system. Due to the large variety of proteins, there is no universal sample rehydration buffer. The sample rehydration buffer must maintain proteins in solution during rehydration of the IPG strips and IEF, and must not have any effect on the pI of the protein. The buffer typically contains a denaturing agent (urea or urea/thiourea), solubilizing agent (non-ionic or zwitterionic detergent and ampholytes), and reducing agent (DTT).
Note: To obtain high resolution, IEF is usually performed under denaturing and reducing conditions. Use a denaturing sample rehydration buffer for sample preparation (see manual for recipe).
How does the ZOOM IPG Runner System work?
The ZOOM IPGRunner System provides a convenient and quick way to perform isoelectric focusing (IEF) of proteins in a vertical mini-gel format using immobilized pH gradient (IPG) strips for two- dimensional (2D) gel electrophoresis.
The major components of the ZOOM IPGRunner System are:
*ZOOM IPGRunner Mini-Cell (Cat. No. ZM0001)
*ZOOM IPGRunner Cassettes (1 pack of 10 cassettes, Cat. No. ZM0003)
*ZOOM Strips (12 ZOOM Strips, pH 3–10 NL, Cat. No. ZM0011)
Using the ZOOM Strips, proteins are separated based on their isoelectric point (pI). The proteins in the sample can be further separated in the second dimension, based on their molecular weight, by placing the IPG Strips into the IPG well of a NuPAGE Bis-Tris ZOOM gel or Tris-Glycine ZOOM gel. The proteins separated in the second dimension can be visualized as spots by the use of SimplyBlue SafeStain or silver staining, or blotted onto membranes. Protein spots can be also excised from the gel or the membranes to be further analyzed by mass spectrometry or chemical microsequencing to facilitate protein identification.
What is the purpose of alkylation prior to IEF?
Alkylation prevents unwanted protein modifications by alkylating cysteines to avoid mixed disulfide formation and reoxidation and this allows for crisper focusing.
Do you still offer ZOOM 2D Protein Solubilizers?
Sorry we have discontinued selling the ZOOM 2D Protein Solubilizers.
What are the main reasons for performing 2D gel electrophoresis of proteins?
The main advantages of performing 2D gel electrophoresis of proteins and applications used for are listed below:
Advantages:
*Simultaneous separation of hundreds to thousands of proteins
*High capacity with superior resolution
*Compatible with further analysis by MS for protein identification and sequencing
Ability to separate and analyze low-abundance proteins
Applications:
*Comparative proteomics: identifying and analyzing differences between complex mixtures of proteins
*Protein profiling, biomarker discovery
*Separation and analysis of protein variants and isoforms
What products do you offer for 2D gel electrophoresis of proteins?
We offer the following products for the first- and second-dimension separation of proteins:
First-dimension separation:
*Vertical gels for separation of proteins based on their isoelectric point (pI) (https://www.thermofisher.com/us/en/home/life-science/protein-expression-and-analysis/protein-gel-electrophoresis/protein-gels/specialized-protein-separation/isoelectric-focusing.html)
*Solution-phase isoelectric focusing of proteins using ZOOM Disks (immobilized buffer disks of specific pH) to reduce sample complexity, enrich low-abundance proteins, and increase the dynamic range of detection (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-gels/specialized-protein-gels/isoelectric-focusing/zoom-ief-fractionator.html)
*Mini gel system for high-throughput isoelectric focusing of proteins using ZOOM IPG (Immobilized pH Gradient) Strips (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-gels/specialized-protein-gels/2d-gel-electrophoresis/zoom-ipgrunner-system.html)
Second-dimension separation:
*ZOOM gels for 2D electrophoresis: NuPAGE Bis-Tris (Cat. No. NP0330BOX) and Tris-Glycine (Cat. No. EC60261BOX) mini gels with IPG wells ( to accommodate 7 cm ZOOM strips) for separation of proteins based on their molecular weight
What is 2D protein gel electrophoresis?
2D protein gel electrophoresis is the separation of proteins in two dimensions. In the first dimension, proteins are separated by their isoelectric point (pI) using isoelectric focusing, and in the second dimension, they are separated by their mass using SDS-PAGE.
For Research Use Only. Not for use in diagnostic procedures.
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.