missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zomepirac Sodium Salt Hydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Zomepirac Sodium Salt Hydrate |
| Molecular Formula | C15H13ClNNaO3 •xH2O |
| Quantity | 500 mg |
| Synonym | 5-(4-Chlorobenzoyl)-1,4-dimethyl-1H-pyrrole-2-acetic Acid Sodium Salt Hydrate, McN 2783-21-98 Hydrate, Sodium Zomepirac Hydrate, Zomax Hydrate, Zomepirac Sodium Hydrate, Zomepirac Sodium Salt Hydrate |
| InChI Formula | InChI=1S/C15H14ClNO3.Na.H2O/c1-9-7-12(8-13(18)19)17(2)14(9)15(20)10-3-5-11(16)6-4-10;;/h3-7H,8H2,1-2H3,(H,18,19);;1H2/q;+1;/p-1 |
| SMILES | O=C(C1=C(C=C(N1C)CC(O[Na])=O)C)C2=CC=C(C=C2)Cl.O |
| IUPAC Name | sodium 2-(5-(4-chlorobenzoyl)-1,4-dimethyl-1H-pyrrol-2-yl)acetate hydrate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?