Learn More
Zolpidem-d6 6-Carboxylic Acid Methyl Ester, TRC
Product Code. 30456148
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456148 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30456148 Supplier Toronto Research Chemicals Supplier No. TRCZ65002710MG
 Restricted Item
Estimated Shipment: 23-10-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Zolpidem-d6 6-Carboxylic Acid Methyl Ester
CAS 1215767-20-6
Molecular Formula C20H15D6N3O3
Quantity 10 mg
Synonym 3-[2-(Dimethylamino)-2-oxoethyl]-2-(4-methylphenyl)-imidazo[1,2-a]pyridine-6-carboxylic Acid-d6 Methyl Ester
InChI Formula InChI=1S/C20H21N3O3/c1-13-5-7-14(8-6-13)19-16(11-18(24)22(2)3)23-12-15(20(25)26-4)9-10-17(23)21-19/h5-10,12H,11H2,1-4H3/i2D3,3D3
SMILES [2H]C([2H])([2H])N(C(=O)Cc1c(nc2ccc(cn12)C(=O)OC)c3ccc(C)cc3)C([2H])([2H])[2H]
IUPAC Name methyl 3-[2-[bis(trideuteriomethyl)amino]-2-oxoethyl]-2-(4-methylphenyl)imidazo[1,2-a]pyridine-6-carboxylate
Molecular Weight (g/mol) 357.44
Formula Weight 357.196