missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconium(IV) Chloride, Sigma-Aldrich™
Description
Zirconium(IV) chloride is a Lewis acid catalyst, widely used in organic synthesis for the preparation of useful key intermediates. Since it is a stable and environmentally friendly catalyst used in various chemical reactions such as carbon-carbon bond formation reactions, reduction reactions, protection, and deprotection reactions.
Specifications
Specifications
| Melting Point | 437°C |
| Quantity | 1 kg |
| MDL Number | MFCD00011306 |
| Synonym | Zirconium(IV) chloride, Zirconium tetrachloride, Zirconium(IV) chloride |
| Molecular Weight (g/mol) | 233.04 |
| Grade | Research |
| Density | 2.80 g/cm3 at 15°C |
| Chemical Name or Material | Zirconium(IV) Chloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?