Learn More
Zirconium(IV) chloride, Sigma-Aldrich™
Description
Zirconium(IV) chloride is a Lewis acid catalyst, widely used in organic synthesis for the preparation of useful key intermediates. Since it is a stable and environmentally friendly catalyst used in various chemical reactions such as carbon-carbon bond formation reactions, reduction reactions, protection, and deprotection reactions.
Specifications
Specifications
| Chemical Name or Material | Zirconium(IV) chloride |
| Melting Point | 437°C |
| CAS | 10026-11-6 |
| Quantity | 5 kg |
| Density | 2.80 g/cm³ at 15°C |
| Synonym | Zirconium(IV) chloride, Zirconium tetrachloride, Zirconium(IV) chloride |
| Molecular Weight (g/mol) | 233.04 |
| Grade | Not Available |