missing translation for 'onlineSavingsMsg'
Learn More

Zinc Oxide, TRC

Product Code. 30010636
Change view
Click to view available options
Quantity:
500 mg
10 g
50 g
Unit Size:
10g
500mg
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010636 50 g 50g
30004028 500 mg 500mg
30003969 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010636 Supplier Toronto Research Chemicals Supplier No. TRCZ43995050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-13-2
Molecular Formula ZnO
Molecular Weight (g/mol) 81.41
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Quantity 50 g
Synonym ActiSorb S 2, Activox C 80, Activox R 50, Actox 14, Actox 16, Actox 216, AdNano ZnO 20, Adnano Z 5, AguaMix 1162, Amalox, Azo-B, Azodox, Bayoxide Z Active, Biocide 3000D, Borchers VP 9802, Bostex 422, Finex;Flowers of zinc, GIAP 10, GinRei, Green Seal 8, Kadox, Kesmon, NanoActive Zinc Oxide, Nanobyk, Nanofine, Nanomax;Nanoshield ZN 3010, Nanotek Z 1021W, Nanotek ZnO, Nanotek ZnO 15, Nanow 2000, Nanox 200, Naoazo, NkS 051, NkS 053, Nyacol DP 5370, Outmine, Oxmer 141, Oxylink 3102, Ozide, Ozlo, PI-KEM, PW 4, PZ, PZ (oxide), Panatetra, Panatetra 0511, Panatetra A, Panatetra B, Panatetra WZ 0501, Panatetra WZ 0511, Panatetra WZ 0531, Panatetra WZ 05F1, Panatetra WZ 501, Panatetra WZ 501L, Parafine ZO, Passtran II 2100, Pazet 23K, Permanent White, Philosopher's wool, Photox 80, Poly-F Powder, Powder base 900, Protox 166, Protox 167, Protox 168, Protox 169, Protox 267, Protox 268, RGT-Z, Red Seal, Red Seal 9, Rhenogran ZnO 85, Ribelin 210, SC 18, SC 18 (oxide), SMZ 017N, SOX 100, SP 500C, SX 8N, SX 8ND, SX 9, SZ 3, Sachtotec, Sato Salbe, Sazex 200, Sazex 2000, Sazex 4000, Shoe-Cleanse, Siloxactif, Snow White, Solasorb UV 200, Super 30, Troysan CP 20, Ultrafine 350, VP 9802, VP AdNano Z 5, VP AdNano Z 805, VP AdNano ZnO 20, VP AdNano ZnO 20DW, Vandem VAC, Vandem VOC, Vandem VPC, W 50, W 50 (oxide), W 9989, WS, WS 55XZ4, WZ 0501, WZ 0531, WZ 501L, WZ 511, White Seal 7, White UV-W 202, White zinc, Whiteseal, ZNAP (oxide), ZNO 01PB, ZNO 02PB, ZNO 04PA, ZNO 04PB, ZNTANB 15E34, ZP 06, ZP 10, ZP 15, ZR 210, ZS 300, ZS 303, ZS 303T, ZW 030A, ZW 143, Zano 20, Zano 30, Zano W 084, Zaobatac Nano 20, Zic Stik 85, Zinc Oxide No. 1, Zinc White 1, Zinc White S, Zinc White Special, Zinc flower, Zinc gelatin, Zinc monoxide, Zinc white, Zinca, Zinca 10, Zinca 20, Zincoid, Zincox Super 1, Zincox Super 10, Zincox Super 30, Zincox Super F 1, Zincox Super F 2, Zincox Super F 2-2 L20, Zincox Super F 3, Zinkoxyd Aktiv, Zinox 350, Zinrex 100R, Zinvisible, Zinvisible Nano-Fine Zinc Oxide;
InChI Formula InChI=1S/O.Zn
Formula Weight 79.9241
Chemical Name or Material Zinc oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.