missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| CAS | 5970-45-6 |
| Color | White |
| Physical Form | Powder |
| Quantity | 500 g |
| Assay Percent Range | 98 to 101% |
| Density | 1.74g/mL |
| Molecular Formula | C4H10O6Zn |
| Synonym | zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn |
| InChI Key | BEAZKUGSCHFXIQ-UHFFFAOYSA-L |
| SMILES | CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2] |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?