Learn More
ZEOprep™ Diol Functionalized
Product Code. 30048965 Shop All Zeochem Products
Change view
Click to view available options
1 kg
100 g
5 kg
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
30048965 100 g
30049026 1 kg
30048949 500 g
30049002 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30048965 Supplier ZEOprep Supplier No. 300186
  • ONLINE EXCLUSIVE PRICE
    250.00 EUR
    229.00 EUR / 100g
    Save 21.00 EUR
    8% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
ZEOprep™ is a packing material providing fast, effective, and reproducible separations. Diol functionalized Irregular shaped silica gel is used in Gravity, Flash, and Medium Pressure Chromatography. The DIOL ligand results in a less polar surface than bare silica.
  • DIOL functionalized for low polarity surface
  • Ideal for lipid applications
  • Suitable for SFC Irregular Amorphous Silica Gel
  • High Purity
  • European Manufacture
  • Swiss accuracy and consistency
  • Excellent batch to batch reproducibility
  • ISO Certification
  • High Chemical and Mechanical Stability
  • Narrow Particles Size Distribution
  • 20 to 500 μm particles size range
  • Available in 60 A or 90 A pore size
  • High loadability
  • Selection of pack sizes available
  • Suitable for gravity, flash, prep and process scale
Specifications
Particle Size 40 to 63 μm
Pore Size 60 Å
Technique Chromatography
Quantity 100 g
Type Silica Gel
Material Silica Gel
Phase DIOL Functionalized
Particle Shape Irregular