missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H12C Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
166.00 EUR - 372.00 EUR
Specifications
| Antigen | ZC3H12C |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18331712
|
Bio-Techne
NBP3-16040-20UL |
20 μg |
166.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18334136
|
Novus Biologicals
NBP3-16040-100UL |
100 μg |
372.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZC3H12C Polyclonal antibody specifically detects ZC3H12C in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| ZC3H12C | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Innate Immunity | |
| PBS, 50% glycerol, pH7.3 | |
| 85463 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| KIAA1726, MCP induced protein 3, MCP-induced protein 3, MCPIP3, probable ribonuclease ZC3H12C, zinc finger CCCH domain-containing protein 12C, zinc finger CCCH-type containing 12C | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 735-883 of human ZC3H12C (NP_203748.1). KAPHLGRSLVATRIDSISDSRLYDSSPSRQRKPYSRQEGLGSWERPGYGIDAYGYRQTYSLPDNSTQPCYEQFTFQSLPEQQEPAWRIPYCGMPQDPPRYQDNREKIYINLCNIFPPDLVRIVMKRNPHMTDAQQLAAAILVEKSQLGY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title