missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ZAP 180013
GREENER_CHOICE

Product Code. 18705756
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18705756

Brand: Tocris Bioscience™ 6821/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Zap70 inhibitor; inhibits interaction with ITAMs

ZAP 180013 is a zeta-chain-associated protein kinase 70 (Zap70) inhibitor (IC50 = 1.8 μM in fluorescence polarization assay). Inhibits the interaction of Zap70 SH2 domain with immunoreceptor tyrosine-based activation motif (ITAMs), within the T-cell antigen receptor (TCR) signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Chloro-N-[(4-chlorophenyl)methyl]-2-(ethylsulfonyl)-N-(2-furanylmethyl)-4-pyrimidinecarboxamide
CAS 873080-25-2
Quantity 10 mg
Molecular Formula C19H17Cl2N3O4S
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.