Learn More
Zaleplon, TRC
Product Code. 30455680
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455680 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455680 Supplier Toronto Research Chemicals Supplier No. TRCZ14500050MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Zaleplon
CAS 151319-34-5
Purity Grade Notes HPLC
Molecular Formula C17 H15 N5 O
Quantity 50 mg
Synonym Zaleplon, Acetamide','N-[3-(3-cyanopyrazolo[1,5-a]pyrimidin-7-yl)phenyl]-N-ethyl-, N-[3-(3-Cyanopyrazolo[1,5-a]pyrimidin-7-yl)phenyl]-N-ethylacetamide, Pyrazolo[1,5-a]pyrimidine','acetamide deriv., CL 284846, L 846, LJC 10846, Siesta, Sonata, Zaleplon, Zerene
InChI Formula InChI=1S/C17H15N5O/c1-3-21(12(2)23)15-6-4-5-13(9-15)16-7-8-19-17-14(10-18)11-20-22(16)17/h4-9,11H,3H2,1-2H3
SMILES CCN(C(=O)C)c1cccc(c1)c2ccnc3c(cnn23)C#N
IUPAC Name N-[3-(3-cyanopyrazolo[1,5-a]pyrimidin-7-yl)phenyl]-N-ethylacetamide
Molecular Weight (g/mol) 305.33
Formula Weight 305.1277
Percent Purity >95
Show More Show Less
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.