Learn More
Zafirlukast, MedChemExpress
Catalog No. 16451866
Encompass_Preferred
Ansicht ändern
Click to view available options
Quantity:
100 mg
500 mg
Packungsgröße:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packungsgröße
16451866 500 mg 500mg
16407336 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Catalog No. 16451866 Lieferant Medchem Express Lieferanten-Nr. HY17492
 Restricted Item
Estimated Shipment: 30-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
MedChemExpress Zafirlukast (ICI 204219) is a potent orally active leukotriene D4 (LTD4) receptor antagonist. Zafirlukast shows anti-asthmatic, anti-inflammatory and anti-bacterial effects.
Zafirlukast (ICI 204219) is a potent orally active leukotriene D4 (LTD4) receptor antagonist. Zafirlukast shows anti-asthmatic, anti-inflammatory and anti-bacterial effects.
Spezifikation
Chemical Name or Material Zafirlukast
Physical Form Powder
CAS 107753-78-6
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C31H33N3O6S
For Use With (Application) COVID-19-immunoregulation
Synonym ICI 204219
Solubility Information DMSO : 100 mg/mL (173.71 mM; Need ultrasonic)
SMILES O=C(OC1CCCC1)NC2=CC3=C(N(C)C=C3CC4=CC=C(C(NS(=O)(C5=CC=CC=C5C)=O)=O)C=C4OC)C=C2
Molecular Weight (g/mol) 575.68
Formula Weight 575.68
Percent Purity 99.1%
Grade Research
Mehr anzeigen Weniger anzeigen