Learn More
Z,Z-Dienestrol Diacetate, TRC
Specifications
Specifications
| Chemical Name or Material | Z,Z-Dienestrol Diacetate |
| CAS | 24705-62-2 |
| Molecular Formula | C22H22O4 |
| Quantity | 100 mg |
| Synonym | beta-Dienestrol diacetate, 4,4'-(Diethylideneethylene)diphenol Diacetate, (Z,Z)-4,4'-(1,2-diethylidene-1,2-ethanediyl)bisphenol Diacetate |
| InChI Formula | InChI=1S/C22H22O4/c1-5-21(17-7-11-19(12-8-17)25-15(3)23)22(6-2)18-9-13-20(14-10-18)26-16(4)24/h5-14H,1-4H3/b21-5-,22-6- |
| SMILES | C\C=C(/C(=C\C)/c1ccc(OC(=O)C)cc1)\c2ccc(OC(=O)C)cc2 |
| IUPAC Name | [4-[(2Z,4Z)-4-(4-acetyloxyphenyl)hexa-2,4-dien-3-yl]phenyl] acetate |
| Molecular Weight (g/mol) | 350.41 |
| Formula Weight | 350.1518 |