missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Z-Val-Glu(OMe)-Ile-Asp(OMe)-Fluoromethylketone, MP Biomedicals™

Product Code. 11484916 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484916 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11484916 Supplier MP Biomedicals™ Supplier No. 03FK01501
  • ONLINE EXCLUSIVE PRICE
    537.00 EUR
    511.35 EUR / 1mg
    Save 25.65 EUR
    4% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Irreversibly inhibits activity of VEID-dependent caspases (e.g., caspase-6). Z-Val-Glu(OMe)-Ile-Asp(OMe)-Fluoromethylketone, MP Biomedicals is a synthetic peptide designed as a methyl ester to facilitate cell permeability. It is used for both in vitro and

If the intended use of this product is on purified or recombinant enzymes, esterase should be added to generate the free carboxyl groups.

Specifications

For Use With (Application) Z-Val-Glu(OMe)-Ile-Asp(OMe)-FMK used for both in vitro and in vivo studies. If the intended use is on purified or recombinant enzymes, esterase should be added to generate the free carboxyl groups.
Content And Storage -20°C
Quantity 1 mg
Molecular Weight (g/mol) 652
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.