missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Z-Tyr-Val-Ala-Asp(OMe)-Fluoromethylketone, MP Biomedicals™

Product Code. 11424916 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424916 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11424916 Supplier MP Biomedicals™ Supplier No. 03FK01301
  • ONLINE EXCLUSIVE PRICE
    498.00 EUR
    473.55 EUR / 1mg
    Save 24.45 EUR
    4% Off
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Irreversibly inhibits activity of Caspase-1 and -4. Z-Tyr-Val-Ala-Asp(OMe)-Fluoromethylketone, MP Biomedicals peptides are potent, cell permeable, irreversible inhibitors that are used for both in vitro and in vivo applications.

Specifications

Content And Storage -20°C
Quantity 1 mg
Molecular Weight (g/mol) 630
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.