Learn More
(Z)-Triprolidine Oxalate Salt, TRC
Specifications
Specifications
| Chemical Name or Material | (Z)-Triprolidine Oxalate Salt |
| CAS | 14343-87-4 |
| Molecular Formula | C19H22N2 . C2H2O4 |
| Quantity | 10 mg |
| Synonym | 2-[(1Z)-1-(4-Methylphenyl)-3-(1-pyrrolidinyl)-1-propenyl]pyridine Oxalate Salt, (Z)-2-[1-(4-Methylphenyl)-3-(1-pyrrolidinyl)-1-propenyl]pyridine Oxalate Salt |
| InChI Formula | InChI=1S/C19H22N2.C2H2O4/c1-16-7-9-17(10-8-16)18(19-6-2-3-12-20-19)11-15-21-13-4-5-14-21;3-1(4)2(5)6/h2-3,6-12H,4-5,13-15H2,1H3;(H,3,4)(H,5,6)/b18-11-; |
| SMILES | Cc1ccc(cc1)\C(=C\CN2CCCC2)\c3ccccn3.OC(=O)C(=O)O |
| IUPAC Name | 2-[(Z)-1-(4-methylphenyl)-3-pyrrolidin-1-ylprop-1-enyl]pyridine;oxalic acid |
| Molecular Weight (g/mol) | 368.4 |
| Formula Weight | 368.1736 |