missing translation for 'onlineSavingsMsg'
Learn More

(Z)-Tetrachlorvinphos, TRC

Product Code. 30422346
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30422346 25 mg
30413678 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30422346 Supplier Toronto Research Chemicals Supplier No. TRCT29230525MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22248-79-9
Molecular Formula C10 H9 Cl4 O4 P
Molecular Weight (g/mol) 363.8992561
Synonym Phosphoric acid, (1Z)-2-chloro-1-(2,4,5-trichlorophenyl)ethenyl dimethyl ester, Phosphoric acid, 2-chloro-1-(2,4,5-trichlorophenyl)ethenyl dimethyl ester, (Z)-, Phosphoric acid, 2-chloro-1-(2,4,5-trichlorophenyl)vinyl dimethyl ester, (Z)- (8CI), (Z)-Tetrachlorovinphos, CVMP, Debantic, Gardicide, Gardona, Rabon, Rabond, SD 8447, Stirofos, Stirophos, Stitifos, Tetrachlorvinfos, Tetrachlorvinphos, Vinfos, cis-Gardona
IUPAC Name [(Z)-2-chloro-1-(2,4,5-trichlorophenyl)ethenyl] dimethyl phosphate
SMILES COP(=O)(OC)O\C(=C/Cl)\c1cc(Cl)c(Cl)cc1Cl

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H9Cl4O4P/c1-16-19(15,17-2)18-10(5-11)6-3-8(13)9(14)4-7(6)12/h3-5H,1-2H3/b10-5-
Formula Weight 363.8993
Chemical Name or Material Tetrachlorvinphos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.