Learn More
Thermo Scientific Chemicals Z-Phe-Ala-fluoromethyl ketone
Description
Cell permeable negative control for FMK-labelled caspase inhibitors. Also an irreversible inhibitor of cathepsins B, L, and S
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 5 mg |
| Description | White to off-white |
| Content And Storage | -30°C to -10°C |
| For Use With (Application) | Cell permeable negative control for FMK-labelled caspase inhibitors. Also an irreversible inhibitor of cathepsins B, L, and S |
| CAS | 105637-38-5 |
| Molecular Formula | C21H23N2O4F, C21H23FN2O4 |
| Formula Weight | 386.42 |
| Synonym | Z-FA-FMK; Z-Phe-Ala-FMK |
| Solubility | Soluble in DMSO or DMF at 20mM |
| Product Type | Z-Phe-Ala-fluoromethyl ketone |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.