Learn More
(Z)-2-(Furan-2-yl)-2-(methoxyimino)acetic Acid, TRC
Product Code. 30455236
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455236 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455236 Supplier Toronto Research Chemicals Supplier No. TRCZ701733250MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (Z)-(Furan-2-yl)(methoxyimino)acetic Acid
CAS 39684-61-2
Molecular Formula C7 H7 N O4
Quantity 250 mg
Synonym (Z)-2-(Furan-2-yl)-2-(methoxyimino)acetic Acid, 2-Furanacetic acid','α-(methoxyimino)-','(αZ)-, 2-Furanacetic acid','α-(methoxyimino)-','(Z)-, (αZ)-α-(Methoxyimino)-2-furanacetic acid, (Z)-2-(2-Furyl)-2-methoxyiminoacetic acid, syn-2-Furyl-2-methoxyiminoacetic acid, (2Z)-2-(2-Furyl)-2-methoxyiminoacetic Acid, (Z)-2-(Furan-2-yl)-2-(methoxyimino)acetic Acid, (Z)-(Furan-2-yl)(methoxyimino)acetic Acid
InChI Formula InChI=1S/C7H7NO4/c1-11-8-6(7(9)10)5-3-2-4-12-5/h2-4H,1H3,(H,9,10)/b8-6-
SMILES CO\N=C(/C(=O)O)\c1occc1
IUPAC Name (2Z)-2-(furan-2-yl)-2-methoxyiminoacetic acid
Molecular Weight (g/mol) 169.135
Formula Weight 169.038