Learn More
(Z)-2-(((1-(2-Aminothiazol-4-yl)-2-((2-hydroxy-2-sulfoethyl)amino)-2-oxoethylidene)amino)oxy)-2-methylpropanoic Acid, TRC
Product Code. 30344483
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30344483 1 mg 1mg
30348168 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30344483 Supplier Toronto Research Chemicals Supplier No. TRCA7334451MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (Z)-2-(((1-(2-Aminothiazol-4-yl)-2-((2-hydroxy-2-sulfoethyl)amino)-2-oxoethylidene)amino)oxy)-2-methylpropanoic Acid
CAS 2519572-35-9
Analyte or Component Names (Z)-2-(((1-(2-Aminothiazol-4-yl)-2-((2-hydroxy-2-sulfoethyl)amino)-2-oxoethylidene)amino)oxy)-2-methylpropanoic Acid
Shipping Condition Room Temperature
Molecular Formula C11H16N4O8S2
InChI Formula InChI=1S/C11H16N4O8S2/c1-11(2,9(18)19)23-15-7(5-4-24-10(12)14-5)8(17)13-3-6(16)25(20,21)22/h4,6,16H,3H2,1-2H3,(H2,12,14)(H,13,17)(H,18,19)(H,20,21,22)/b15-7-
Solution Type Neat
SMILES NC1=NC(/C(C(NCC(O)S(=O)(O)=O)=O)=N/OC(C)(C)C(O)=O)=CS1
IUPAC Name (Z)-2-(((1-(2-aminothiazol-4-yl)-2-((2-hydroxy-2-sulfoethyl)amino)-2-oxoethylidene)amino)oxy)-2-methylpropanoic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 396.4
Quantity 1 mg
Formula Weight 396.04