missing translation for 'onlineSavingsMsg'
Learn More

Z-12-Pentacosene (>85%), TRC

Product Code. 30425577
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30425577 5 mg -
30414568 10 mg -
30413629 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30425577 Supplier Toronto Research Chemicals Supplier No. TRCP5121505MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56927-05-0
Molecular Formula C25H50
Molecular Weight (g/mol) 350.68
Synonym (12Z)-12-Pentacosene, (Z)-12-Pentacosene, (Z)-Pentacos-12-ene
IUPAC Name (Z)-pentacos-12-ene
SMILES CCCCCCCCCCCC/C=C\CCCCCCCCCCC

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C25H50/c1-3-5-7-9-11-13-15-17-19-21-23-25-24-22-20-18-16-14-12-10-8-6-4-2/h23,25H,3-22,24H2,1-2H3/b25-23-
Formula Weight 350.3913
Chemical Name or Material Z-12-Pentacosene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.