Learn More
Yunnaconitine, TRC
Product Code. 30454932
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454932 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30454932 Supplier Toronto Research Chemicals Supplier No. TRCY10090010MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Yunnaconitine
CAS 70578-24-4
Molecular Formula C35 H49 N O11
Quantity 10 mg
Synonym (1α,3α,6α,14α,16β)-20-Ethyl-1,6,16-trimethoxy-4-(methoxymethyl)-8-acetate 14-(4-methoxybenzoate)aconitane-3,8,13,14-tetrol, Isoaconitine (6CI), Vilmorrianine B (7CI), 11aH-12,3,6a-Ethanylylidene-7,9-methanonaphth[2,3-b]azocine','Aconitane-3,8,13,14-tetrol Deriv., 3α,13-Dihydroxyforesaconitine, Guayewuanine B, Yunaconitine
InChI Formula InChI=1S/C35H49NO11/c1-8-36-16-32(17-41-3)22(38)13-23(43-5)35-21-14-33(40)24(44-6)15-34(47-18(2)37,26(29(35)36)27(45-7)28(32)35)25(21)30(33)46-31(39)19-9-11-20(42-4)12-10-19/h9-12,21-30,38,40H,8,13-17H2,1-7H3/t21-,22-,23+,24+,25-,26+,27+,28-,29-,30-,32+,33+,34-,35+/m1/s1
SMILES CCN1C[C@]2(COC)[C@H](O)C[C@H](OC)[C@@]34[C@@H]5C[C@]6(O)[C@H](C[C@](OC(=O)C)([C@@H]([C@H](OC)[C@H]23)[C@@H]14)[C@H]5[C@H]6OC(=O)c7ccc(OC)cc7)OC
Molecular Weight (g/mol) 659.76
Formula Weight 659.3306