Learn More
YM-90709, MedChemExpress
Product Code. 16493716
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493716 25 mg 25mg
16477988 10 mg 10mg
16443917 50 mg 50mg
16420458 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16493716 Supplier Medchem Express Supplier No. HY19969
MedChemExpress YM-90709 is a novel IL-5 inhibitor which selectively blocks the binding of IL-5 to the IL-5 receptor (IL-5R).YM-90709 potently inhibits the binding of [125I]-IL-5 to IL-5R on human peripheral eosinophils and eosinophilic HL-60 clone 15 cells with IC50 values of 1.0 and 0.57 μM.
YM-90709 is a novel IL-5 inhibitor which selectively blocks the binding of IL-5 to the IL-5 receptor (IL-5R).YM-90709 potently inhibits the binding of [125I]-IL-5 to IL-5R on human peripheral eosinophils and eosinophilic HL-60 clone 15 cells with IC50 values of 1.0 and 0.57 μM.
Specifications
Chemical Name or Material YM-90709
CAS 163769-88-8
Color Orange
Purity Grade Notes Research
Molecular Formula C22H21N3O2
Quantity 25 mg
Solubility Information DMSO : 62.5 mg/mL (173.89 mM; Need ultrasonic)
SMILES CC(CC1=CC(OC)=C(OC)C=C12)(C)N3C2=CC4=NC5=CC=CC=C5N=C43
Molecular Weight (g/mol) 359.42
Formula Weight 359.42
Percent Purity 99.52%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation